Hydroquinon
Glass bottle for solids, 1 kg
Katalog # ROTH3586.3
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: 1,4-Dihydroxybenzen, Benzen-1,4-diol
- CAS nummer:123-31-9
- Formel:C₆H₄(OH)₂
- MDL nummer:MFCD00002339
- Molecular Weight:110.11 g/mol
- Merck Index:13,04833
- Kogende Pt:285 °C (1013 hPa)
- Smeltende Pt:172 °C
- Tæthed:1,332 g/cm³ (20 °C)
- Flash Pt:165 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-617-8
- FN:3077
- ADR:9,III
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg