Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Diethylamin

Diethylamin

Glass bottle, 1 L
Catalog # ROTH3495.2
Flammable
Corrosive
Irritant
Marketsource
Supplier:   Roth Carl
Diethylamin
Catalog # ROTH3495.2
Supplier:  Roth Carl
Marketsource
CAS Number:  109-89-7
Diethylamin
Diethylamin

Glass bottle, 1 L

CAS Number:  109-89-7
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:109-89-7
  • Formula:(C₂H₅)₂NH
  • MDL Number:MFCD00009032
  • Molecular Weight:73.14 g/mol
  • Merck Index:13,03138
  • Boiling Pt:55,5 °C (1013 hPa)
  • Melting Pt:–50 °C
  • Density:0,7048 g/cm³ (20 °C)
  • Flash Pt:–39 °C
  • Storage Temperature:Stue temperatur
  • EINECS:203-716-3
  • UN:1154
  • ADR:3,II
Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.