Diethylamin
Glass bottle, 250 mL
ROTH3495.1
Marketsource
- :109-89-7
- :(C₂H₅)₂NH
- :MFCD00009032
- :73.14 g/mol
- :13,03138
- :55,5 °C (1013 hPa)
- :–50 °C
- :0,7048 g/cm³ (20 °C)
- :–39 °C
- :Stue temperatur
- :203-716-3
- :1154
- :3,II
- Pakning:Glass bottle
- Pakn:250 mL