Order Entry
Denmark
Orders LinkContactUsLinkComponent
L(+)-Cystein

L(+)-Cystein

Glass bottle for solids, 10 g
Katalog # ROTH3467.1
Irriterende
Marketsource
Leverandør:   Roth Carl
L(+)-Cystein
Katalog # ROTH3467.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  52-90-4
L(+)-Cystein

Glass bottle for solids, 10 g

CAS nummer:  52-90-4
Price
141,18 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents
Synonymer:  H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
  • CAS nummer:52-90-4
  • Formel:HSCH₂CH(NH₂)CO₂H
  • MDL nummer:MFCD00064306
  • Molecular Weight:121.16 g/mol
  • Merck Index:13,02810
  • Kogende Pt:254 °C (1013 hPa)
  • Smeltende Pt:115 °C
  • Tæthed:1,523 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-158-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.