(±)-1-Methoxy-2-propanol
Glass bottle, 1 L
ROTH3123.1
Marketsource
: (±)-Monopropylene glycol methyl ether, 1-Methoxy-2-propanol
- :107-98-2
- :CH₃CH(OH)CH₂OCH₃
- :MFCD00004537
- :90.12 g/mol
- :119,6 °C (1013 hPa)
- :–97 °C
- :0,92 g/cm³ (20 °C)
- :31 °C
- :Stue temperatur
- :203-539-1
- :3092
- :3,III
- Pakning:Glass bottle
- Pakn:1 L