L-Alanin
Glass bottle for solids, 25 g
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
- CAS Number:56-41-7
- Formula:C₃H₇NO₂
- MDL Number:MFCD00064410
- Molecular Weight:89.09 g/mol
- Merck Index:13,00203
- Boiling Pt:189 °C (1013 hPa)
- Melting Pt:79 °C
- Density:1,4 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-273-8
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:25 g



