Natriumdodecylsulfat (SDS)
Glass bottle for solids, 500 g
Katalog # ROTH2326.3
Marketsource
Leverandør: Roth Carl
Natriumdodecylsulfat (SDS)
Katalog # ROTH2326.3
Leverandør: Roth Carl
Marketsource
CAS nummer: 151-21-3
Gå til:
Product Details & Documents
Synonymer: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
- CAS nummer:151-21-3
- Formel:CH₃(CH₂)₁₁OSO₃Na
- MDL nummer:MFCD00036175
- Molecular Weight:288.38 g/mol
- Merck Index:14,08636
- Smeltende Pt:204…207 °C
- Tæthed:1,1 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:205-788-1
- FN:1325
- ADR:4.1,II
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



