X-Gal
Glass bottle for solids, 5 g
ROTH2315.4
Marketsource
: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
- :7240-90-6
- :C₁₄H₁₅BrClNO₆
- :MFCD00005666
- :408.63 g/mol
- :237…239 °C
- :Fryser
- :230-640-8
- Pakning:Glass bottle for solids
- Pakn:5 g