Orders LinkContactUsLinkComponent
X-Gal
Glass bottle for solids, 5 g
  ROTH2315.4
Marketsource
 :   Roth Carl
X-Gal
  ROTH2315.4
 :  Roth Carl
Marketsource
 :  7240-90-6
X-Gal
Glass bottle for solids, 5 g
 :  7240-90-6
1 * 5 g
 :  BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
  •  :7240-90-6
  •  :C₁₄H₁₅BrClNO₆
  •  :MFCD00005666
  •  :408.63 g/mol
  •  :237…239 °C
  •  :Fryser
  •  :230-640-8
  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g