Acetonitril
Glass bottle, 2,5 L
Katalog # ROTH2216.2
Marketsource
Leverandør: Roth Carl
Acetonitril
Katalog # ROTH2216.2
Leverandør: Roth Carl
Marketsource
CAS nummer: 75-05-8
Gå til:
Product Details & Documents
Synonymer: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- CAS nummer:75-05-8
- Formel:H₃CCN
- MDL nummer:MFCD00001878
- Molecular Weight:41.05 g/mol
- Merck Index:13,00071
- Kogende Pt:81,6 °C (1013 hPa)
- Smeltende Pt:–45,7 °C
- Tæthed:0,782 g/cm³ (20 °C)
- Flash Pt:2 °C (closed cup)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-835-2
- FN:1648
- ADR:3,II
- NÅ:01-2119471307-38
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
- Cat. No.:ROTH2216.2
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø



