Trypsin
Glass bottle for solids, 100 mg
Katalog # ROTH2193.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 mg
- Cat. No.:ROTH2193.1
- CAS-nr:9002-07-7
- MDL No.:MFCD00082094
- Renhed:2X crystallized
- Opbevaring betingelser:Refrigerator
- Enzym navn:Trypsin