Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Push-in stoppers for tubes

Push-in stoppers for tubes

gul, 17 mm
Katalog # SIMPT401-10Y
Leverandør:   Simport Scientific
Push-in stoppers for tubes
Katalog # SIMPT401-10Y
Leverandør:  Simport Scientific
Push-in stoppers for tubes
Push-in stoppers for tubes

gul, 17 mm

Price
736,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents

HDPE. Fit tubes of 15 ml capacity and 17 mm Ø.

  • Impermeable to liquids
  • Can be inserted in tubes and removed again easily with one hand
  • Economical, can also be used as disposable stoppers
Specifikationer
  • Ø bund:
    17 mm
  • Farve:
    gul
  • Seq:
    16
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.