Orders LinkContactUsLinkComponent
Staining dishes, WHEATON®
Staining dish complete, includes dish, cover and rack, 185×88×68 mm
Catalog # 720-1031
Supplier:   DWK Life Sciences
Staining dishes, WHEATON®
Catalog # 720-1031
Supplier:  DWK Life Sciences
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Staining dish complete, includes dish, cover and rack, 185×88×68 mm
Price
17.680,00 kr
Log in for availability
1 * 6 Items
Product Details & Documents
Soda-lime glass and stainless steel.
  • Handle permanently attached
  • Hinged closure
  • Easy insertion and removal of slides
The slotted rack hold 50 microscope slides 75×25 mm, 75×38 mm or 75×51 mm. Rack is made of non tarnishable, stainless steel that is resistant to staining solutions.
Specifications
  • Beskrivelse:
    Staining dish complete, includes dish, cover and rack
  • B×D×H:
    185×88×68 mm