Lakmus
Glass bottle for solids, 100 g
Katalog # CHRO1A-016-100
Marketsource
Leverandør: CHROMA
Gå til:
Product Details & Documents
- CAS nummer:1393-92-6
- MDL nummer:MFCD00131528
- EINECS:215-739-6
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



