Liner seals
Graphite liner seals, for Bruker/Varian 1078/1079 GCs, 450 °C
Katalog # REST22683
Leverandør: Restek
Gå til:
Product Details & Documents
Specifikationer
- Temperaturområde:450 °C
- Beskrivelse:Graphite liner seals, for Bruker/Varian 1078/1079 GCs
- Seq:01