Dropping funnels, conical shape, without pressure balance
2000 ml, 29/32 NS, 29/32 NS
532-0051
Borosilicate glass 3.3.
- Screw thread retaining device
- With inscription label, without graduation
- Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and stopcock with glass or PTFE plug.
- Konusstr.:29/32 NS
- Kappestr.:29/32 NS
- Kapacitet:2000 ml