Reaction vessels, cylindrical
4000 ml, laboratory flange LF 150, 160 mm, 325 mm
LENZ06000479
DURAN®, borosilicate glass, clear, with round bottom.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With easily readable graduation and laboratory flat flange (LF) with groove.
- Udv.Ø:160 mm
- med:laboratory flange LF 150
- Kapacitet:4000 ml
- Højde:325 mm
- Seq:11