Orders LinkContactUsLinkComponent
Counting Chambers, Neubauer
Counting Chambers, Neubauer
Counting chamber with double ruling, Neubauer, 0,1 mm
Katalog # 631-0925
Counting Chambers, Neubauer
Katalog # 631-0925
Counting Chambers, Neubauer
Counting Chambers, Neubauer
Counting chamber with double ruling, Neubauer, 0,1 mm
Price
417,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These counting chambers are designed for blood tests and urine analysis.
  • Counting erythrocytes corresponds exactly to Thoma method
  • Counting leucocytes in five large squares
  • Evaluation just as indicated for Thoma ruling
Consisting of nine large squares – The central square shows the Thoma ruling and the other eight squares are subdivided.
Documents
Specifikationer
  • Kammerdybde:
    0,1 mm
  • Beskrivelse:
    Counting chamber with double ruling, Neubauer
  • Seq:
    02