Heating tapes, IT-SiS10
Heating tape IT-SiS10, 2 m length, 200 W
Katalog # ISOP162874-000
Leverandør: ISOPAD
Gå til:
Product Details & Documents
Isopad IT-SiS10 is a preterminated, flexible, and waterproof silicone insulated heating tape. Four silicone insulated heating conductors are embedded in a protective overbraid made of cupro-nickel. The temperature depends on the power and application.
- Maximum operating temperature: 200 °C
- Width×Thickness: 11×5 mm (±10%)
- Maximum power per meter 100 W/m
It is non hazardous and used in ordinary areas. It is used for temperature modification of pipes and structures in industries and laboratories.
Specifikationer
- Beskrivelse:Heating tape IT-SiS10, 2 m length, 200 W
- Seq:02



