Graphite sealing rings
Graphite sealing ring, 2 pack
Katalog # REST21899
Leverandør: Restek
Gå til:
Product Details & Documents
These graphite sealing rings are suitable for Thermo Scientific TRACE, 8000, 8000 TOP and Focus SSL instruments. They fit liners with 8 mm outer diameter.
Specifikationer
- Beskrivelse:Graphite sealing ring, 2 pack
- Seq:02