Orders LinkContactUsLinkComponent
Glass stoppers with hexagonal grip
Glass stoppers with hexagonal grip
24/29, Brunt glas
Katalog # WITG0.303.024
Leverandør:   witeg Labortechnik
Glass stoppers with hexagonal grip
Katalog # WITG0.303.024
Leverandør:  witeg Labortechnik
Glass stoppers with hexagonal grip
Glass stoppers with hexagonal grip
24/29, Brunt glas
Price
65,70 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Hollow standard taper stoppers with hexagonal grip, made of borosilicate glass 3.3.
  • Acc. to DIN 12252
Specifikationer
  • NS:
    24/29
  • Model:
    Brunt glas