Glass stoppers with hexagonal grip
24/29, Brunt glas
Katalog # WITG0.303.024
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Hollow standard taper stoppers with hexagonal grip, made of borosilicate glass 3.3.
- Acc. to DIN 12252
Specifikationer
- NS:24/29
- Model:Brunt glas