Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Skin blocks

Skin blocks

Block of skin, SOMSO-Plast®, 1,3 kg, 210×200×110 mm
Katalog # SOMMKS4
Leverandør:   SOMMER MARCUS
Skin blocks
Katalog # SOMMKS4
Leverandør:  SOMMER MARCUS
Skin blocks
Skin blocks

Block of skin, SOMSO-Plast®, 1,3 kg, 210×200×110 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

One piece on a green base enlarged 70 times with:

  • a - Scalp with hair
  • b - Axilla
  • c - Hairless skin of the sole of the foot
Specifikationer
  • Vægt:
    1,3 kg
  • Beskrivelse:
    Block of skin
  • Materiale:
    SOMSO-Plast®
  • B×D×H:
    210×200×110 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.