Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Ordreindtastning
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Oxygen, dissolved - CHEMets® Refill 1 * 1 Each
Oxygen, dissolved - CHEMets® Refill 1 * 1 Each
Print
Oxygen, dissolved - CHEMets® Refill 1 * 1 Each
Catalog # CHETR-7501
Supplier:
CHEMetrics
Oxygen, dissolved - CHEMets® Refill 1 * 1 Each
Catalog # CHETR-7501
Supplier:
CHEMetrics
CAS Number:
Expand image
Return to Top
Oxygen, dissolved - CHEMets® Refill 1 * 1 Each
Add to cart
CAS Number:
Price
807,00 kr
Log in
for availability
Unit of Measure
1 * 1 Each
-
+
Add to cart
Return Policy
Product Details & Documents