Orders LinkContactUsLinkComponent
Wiring
Wiring
Ledning, 0,5 mm, Nikkel-krom
Catalog # 761-0148
Marketsource
Wiring
Catalog # 761-0148
Marketsource
Wiring
Wiring
Ledning, 0,5 mm, Nikkel-krom
Price
Inquire For Price
Log in for availability
1 * 50 m
Product Details & Documents
Bare wire in a roll, on a plastic former.
Specifications
  • Beskrivelse:
    Ledning, 0,5 mm
  • Materiale:
    Nikkel-krom