Orders LinkContactUsLinkComponent
Engine models
Stirling engine
  765-0172
Marketsource
Engine models
  765-0172
Marketsource
Engine models
Engine models
Stirling engine
1 * 1 Items
Intended for use on overhead projectors for demonstration of function and principle for different types of motors. Made of strong, transparent plastic. Equipped with suction feet.
  • Dimensions: 250 × 250 mm
  • Beskrivelse:
    Stirling engine