Engine models
Otto engine, two-stroke
Katalog # 765-0174
Gå til:
Discontinued
Product Details & Documents
Intended for use on overhead projectors for demonstration of function and principle for different types of motors. Made of strong, transparent plastic. Equipped with suction feet.
- Dimensions: 250 × 250 mm
Specifikationer
- Beskrivelse:Otto engine, two-stroke