Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bromcresol grønt

Bromcresol grønt

Glass bottle, 250 mL
Katalog # CHRO3L-082-0250
Marketsource
Leverandør:   CHROMA
Bromcresol grønt
Katalog # CHRO3L-082-0250
Leverandør:  CHROMA
Marketsource
CAS nummer:  76-60-8
Bromcresol grønt
Bromcresol grønt

Glass bottle, 250 mL

CAS nummer:  76-60-8
Price
Spørg for pris
Log ind for tilgængelighed
1 * 250 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Synonymer:  3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS nummer:76-60-8
  • Formel:C₂₁H₁₄Br₄O₅S
  • MDL nummer:MFCD00005874
  • Molecular Weight:698.02 g/mol
  • Merck Index:13,01370
  • Smeltende Pt:217…218 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-972-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    250 mL

Specifikationer