Bromcresol grønt
Glass bottle, 100 mL
Catalog # CHRO3L-082-0100
Marketsource
Supplier: CHROMA
Jump to:
Product Details & Documents
Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-60-8
- Formula:C₂₁H₁₄Br₄O₅S
- MDL Number:MFCD00005874
- Molecular Weight:698.02 g/mol
- Merck Index:13,01370
- Melting Pt:217…218 °C
- Storage Temperature:Stue temperatur
- EINECS:200-972-8
Specifications
Specifications
- Pakning:Glass bottle
- Pakn:100 mL



