Moisture control by-pass lines
Moisture control by-pass line for Tekmar 3000
Katalog # REST21035
Leverandør: Restek
Gå til:
Product Details & Documents
These moisture control by-pass lines for Tekmar instruments are suitable for U.S EPA methods 8260, 524.2 and OLM4.1.
- Silcosteel®-deactivated tubing for increased inertness
- Increases response for ketones, alcohols and acetates
- Easy attachment in minutes
Specifikationer
- Beskrivelse:Moisture control by-pass line for Tekmar 3000