Orders LinkContactUsLinkComponent
Moisture control by-pass lines
Moisture control by-pass lines
Moisture control by-pass line for Tekmar 3000
Katalog # REST21035
Leverandør:   Restek
Moisture control by-pass lines
Katalog # REST21035
Leverandør:  Restek
Moisture control by-pass lines
Moisture control by-pass lines
Moisture control by-pass line for Tekmar 3000
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These moisture control by-pass lines for Tekmar instruments are suitable for U.S EPA methods 8260, 524.2 and OLM4.1.
  • Silcosteel®-deactivated tubing for increased inertness
  • Increases response for ketones, alcohols and acetates
  • Easy attachment in minutes
Specifikationer
  • Beskrivelse:
    Moisture control by-pass line for Tekmar 3000