Orders LinkContactUsLinkComponent
Trypsinising flasks
Trypsinising flasks
5000 ml
Katalog # WITG5.507.308
Leverandør:   witeg Labortechnik
Trypsinising flasks
Katalog # WITG5.507.308
Leverandør:  witeg Labortechnik
Trypsinising flasks
Trypsinising flasks
5000 ml
Price
2.680,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Trypsinising flasks, graduated, designed for converting tissue fragments and homogenates into cell suspensions by trypsin digestion of connective proteins. Flutes in flask enhance vigorous agitation.
Specifikationer
  • Kapacitet:
    5000 ml
  • Seq:
    06