Standard ground joints
- NS, 24/29 NS, 28 mm
Katalog # QUTW1032
Leverandør: QUARTZTECHNISCHE WERKSTATTEN
Gå til:
Product Details & Documents
Connections, quartz glass, with standard ground joint according to DIN 12 249.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving high resistance to temperature changes.
Specifikationer
- Konusstr.:- NS
- Kappestr.:24/29 NS
- Rør udv. Ø:28 mm
- Seq:03