Replacement inlet seals
Inlet seal, used with 0,53 mm ID dual column, stainless steel, 1,6 mm Ø int.
Katalog # REST20393
Leverandør: Restek
Gå til:
Product Details & Documents
Inlet seals are used at the base of the Agilent 5890/6890/7890 GC injection port, which contacts the sample and should be changed frequently to prevent adsorption of active compounds. In addition, septum fragments and sample residue accumulate on the disk surface, requiring disk replacement.
Specifikationer
- Beskrivelse:Inlet seal, used with 0,53 mm ID dual column, stainless steel, 1,6 mm Ø int.
- Seq:14
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement inlet seal washers