Orders LinkContactUsLinkComponent
Replacement inlet seals
Replacement inlet seals
Inlet seal, used with 0,53 mm ID dual column, stainless steel, 1,6 mm Ø int.
Catalog # REST20392
Supplier:   Restek
Replacement inlet seals
Catalog # REST20392
Supplier:  Restek
Replacement inlet seals
Replacement inlet seals
Inlet seal, used with 0,53 mm ID dual column, stainless steel, 1,6 mm Ø int.
Price
Inquire For Price
Log in for availability
1 * 2 Items
Product Details & Documents
Inlet seals are used at the base of the Agilent 5890/6890/7890 GC injection port, which contacts the sample and should be changed frequently to prevent adsorption of active compounds. In addition, septum fragments and sample residue accumulate on the disk surface, requiring disk replacement.
Specifications
  • Beskrivelse:
    Inlet seal, used with 0,53 mm ID dual column, stainless steel, 1,6 mm Ø int.
  • Seq:
    13
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • REST21710
    Replacement inlet seal washers
    Log in for availability
    1 * 15 Items - 127,00 kr