Orders LinkContactUsLinkComponent
Retord rod, without thread
Retord rod, without thread
13 mm, 915 mm
Katalog # 241-7871
Leverandør:   R L ENTERPRISES
Retord rod, without thread
Katalog # 241-7871
Leverandør:  R L ENTERPRISES
Retord rod, without thread
Retord rod, without thread
13 mm, 915 mm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Anodized aluminium.
Ø = 13 mm
Specifikationer
  • Ø:
    13 mm
  • Længde:
    915 mm