Orders LinkContactUsLinkComponent
Distilling bridges
Distilling bridges
200 mm, 19/26 NS
Katalog # WITG1.200.200
Leverandør:   witeg Labortechnik
Distilling bridges
Katalog # WITG1.200.200
Leverandør:  witeg Labortechnik
Distilling bridges
Distilling bridges
200 mm, 19/26 NS
Price
548,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling links without condenser, with two vertical NS cones.
Specifikationer
  • Konusstr.:
    19/26 NS
  • Kondenserlængde:
    200 mm