Orders LinkContactUsLinkComponent
Fractionating columns, Vigreux
Fractionating columns, Vigreux
600 mm, 29/32 NS, 29/32 NS
Catalog # 201-0179
Supplier:   witeg Labortechnik
Fractionating columns, Vigreux
Catalog # 201-0179
Supplier:  witeg Labortechnik
Fractionating columns, Vigreux
Fractionating columns, Vigreux
600 mm, 29/32 NS, 29/32 NS
Price
1.520,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With or without glass jacket.
Specifications
  • Konusstr.:
    29/32 NS
  • Kappestr.:
    29/32 NS
  • Samlet længde:
    600 mm