Orders LinkContactUsLinkComponent
Distilling heads
19/26 NS, 29/32 NS
  201-0207
Distilling heads
  201-0207
Distilling heads
Distilling heads
19/26 NS, 29/32 NS
412,00 kr
1 * 1 Items
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With one NS socket and two NS cones, descending angle: 75°.
  • Konus til kondenser:
    19/26 NS
  • Konus til kolbe:
    29/32 NS