Distilling bridges
300 mm, 24/29 NS
Katalog # WITG1.200.300
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3,3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling links without condenser, with two vertical NS cones.
Specifikationer
- Konusstr.:24/29 NS
- Kondenserlængde:300 mm