Condensers, Graham
250 mm, 29/32 NS, 29/32 NS
Katalog # WITG1.738.250
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3,3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Coil type, Graham pattern, suitable for either reflux or distillation work. Water connection with glass olives.
Specifikationer
- Konusstr.:29/32 NS
- Kondenserlængde:250 mm
- Kappestr.:29/32 NS