Orders LinkContactUsLinkComponent
Stoppers for conical ground joints
Stoppers for conical ground joints
12/21, Clear glass
Katalog # 217-9096
Leverandør:   Lenz Laborglas
Stoppers for conical ground joints
Katalog # 217-9096
Leverandør:  Lenz Laborglas
Stoppers for conical ground joints
Stoppers for conical ground joints
12/21, Clear glass
Price
88,60 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass stopper, clear, solid or semi hollow.
  • Excellent chemical resistance
  • High thermal resistance and minimal heat expansion results in high resistance to temperature changes
  • Octagonal top allows easy release and removal of the stopper
Certifications: DIN 12252
Documents
Specifikationer
  • NS:
    12/21
  • Model:
    Clear glass