Stoppers for conical ground joints
12/21, Clear glass
Katalog # 217-9096
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilicate glass stopper, clear, solid or semi hollow.
- Excellent chemical resistance
- High thermal resistance and minimal heat expansion results in high resistance to temperature changes
- Octagonal top allows easy release and removal of the stopper
Certifications: DIN 12252
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- NS:12/21
- Model:Clear glass