Orders LinkContactUsLinkComponent
Butyrometer original Gerber
Butyrometer original Gerber
Butyrometer til ost, 0-40 %:0,5, ISO/DIN/NE 239 - 40
Katalog # 709-0134
Leverandør:   Gerber Instruments AG
Butyrometer original Gerber
Katalog # 709-0134
Leverandør:  Gerber Instruments AG
Butyrometer original Gerber
Butyrometer til ost, 0-40 %:0,5, ISO/DIN/NE 239 - 40
Price
694,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass.
Butyrometers are used to determine the fat content of dairy samples. The product is treated with concentrated sulphuric acid to disrupt the emulsion. The resulting mixture is centrifuged in the butyrometer and the fat content read directly off the scale.
Specifikationer
  • Beskrivelse:
    Butyrometer til ost, 0-40 %:0,5, ISO/DIN/NE 239 - 40