Butyrometer original Gerber
Butyrometer til mælk, 0 - 8 %:0,1, ISO 510 - 8
709-0133
DURAN®, borosilicate glass.
Butyrometers are used to determine the fat content of dairy samples. The product is treated with concentrated sulphuric acid to disrupt the emulsion. The resulting mixture is centrifuged in the butyrometer and the fat content read directly off the scale.
- Beskrivelse:Butyrometer til mælk, 0 - 8 %:0,1, ISO 510 - 8