Orders LinkContactUsLinkComponent
Glass wool
Glass wool
Base-deactivated glass wool, ideal for amines and other basic compounds
Catalog # REST20999
Supplier:   Restek
Glass wool
Catalog # REST20999
Supplier:  Restek
Glass wool
Glass wool
Base-deactivated glass wool, ideal for amines and other basic compounds
Price
590,00 kr
Log in for availability
1 * 10 g
Product Details & Documents
Documents
Specifications
  • Beskrivelse:
    Base-deactivated glass wool, ideal for amines and other basic compounds
  • Seq:
    03