Fingertight capillary column nuts
Finger-tight capillary column nut, for use with standard 1,59 mm-type ferrules
Katalog # REST21312
Leverandør: Restek
Gå til:
Product Details & Documents
These fingertight capillary column nuts rapidly tighten without wrenches and can be used with 0,25, 0,32, or 0,53 mm ID columns.
Specifikationer
- Beskrivelse:Finger-tight capillary column nut, for use with standard 1,59 mm-type ferrules
- Seq:02