Sensors
Oxygen (DT222A), DO2: 0 - 12,5 mg/L
O2: 0 - 25 %
O2: 0 - 25 %
Katalog # 765-0635
Leverandør: FOURIER SYSTEMS
Gå til:
Product Details & Documents
Approximately 65 sensors available for MultiLogPRO/Nova 5000/USBLink. See below for examples. Please visit www.fourier-sys.com for more detailed information.
Specifikationer
- Beskrivelse:Oxygen (DT222A)
- Seq:13
- Område:DO2: 0 - 12,5 mg/L
O2: 0 - 25 %