Orders LinkContactUsLinkComponent
Sensors
Magnetic field (DT156), ±10 mT; ±0,2 mT
Katalog # 765-0634
Leverandør:   FOURIER SYSTEMS
Sensors
Katalog # 765-0634
Leverandør:  FOURIER SYSTEMS
Sensors
Sensors
Magnetic field (DT156), ±10 mT; ±0,2 mT
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Approximately 65 sensors available for MultiLogPRO/Nova 5000/USBLink. See below for examples. Please visit www.fourier-sys.com for more detailed information.
Specifikationer
  • Beskrivelse:
    Magnetic field (DT156)
  • Seq:
    12
  • Område:
    ±10 mT; ±0,2 mT