Sensors
Magnetic field (DT156), ±10 mT; ±0,2 mT
Katalog # 765-0634
Leverandør: FOURIER SYSTEMS
Gå til:
Product Details & Documents
Approximately 65 sensors available for MultiLogPRO/Nova 5000/USBLink. See below for examples. Please visit www.fourier-sys.com for more detailed information.
Specifikationer
- Beskrivelse:Magnetic field (DT156)
- Seq:12
- Område:±10 mT; ±0,2 mT