Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sensors

Sensors

Sensor for acceleration, DT138, ±5 g (±49 m/s²)
Katalog # 765-0633
Leverandør:   FOURIER SYSTEMS
Sensors
Katalog # 765-0633
Leverandør:  FOURIER SYSTEMS
Sensors
Sensors

Sensor for acceleration, DT138, ±5 g (±49 m/s²)

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Approximately 65 sensors available for MultiLogPRO/Nova 5000/USBLink. See below for examples. Please visit www.fourier-sys.com for more detailed information.

Specifikationer
  • Beskrivelse:
    Sensor for acceleration, DT138
  • Seq:
    18
  • Område:
    ±5 g (±49 m/s²)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.