Bacteria prepared slides
Bacteria microscope slides
Katalog # 763-0361
Leverandør: LIEDER JOHANNES
Gå til:
Product Details & Documents
This bacteriological slide includes.
- Streptococcus
- Diphtheria bacilli
- Spirillum
- Staphylococcus
- Oral flora
- Septic arthritis
- Syphilis
- Tuberculosis (Tb)
- Tetanus germs
- Coliform
- Salmonella germs
- Typhoid fever germs
Leveringsinformation: Supplied with pictures and explanatory texts.
Specifikationer
- Beskrivelse:Bacteria microscope slides