Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Skin blocks

Skin blocks

Block of skin, SOMSO-Plast®, 2,2 kg, 250×470×150 mm
Katalog # SOMMKS3
Leverandør:   SOMMER MARCUS
Skin blocks
Katalog # SOMMKS3
Leverandør:  SOMMER MARCUS
Skin blocks
Skin blocks

Block of skin, SOMSO-Plast®, 2,2 kg, 250×470×150 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

One piece on a green base enlarged 70 times with:

  • a - Scalp with hair
  • b - Axilla
  • c - Hairless skin of the sole of the foot
Specifikationer
  • Vægt:
    2,2 kg
  • Beskrivelse:
    Block of skin
  • Materiale:
    SOMSO-Plast®
  • B×D×H:
    250×470×150 mm