Gravity hydrometers
ASTM hydrometer, 19…31 °API, 330 mm
Catalog # AMARH825004
Supplier: Amarell
Jump to:
Product Details & Documents
These gravity hydrometers are designed with - and without - thermometer.
- Paper scale
- Type designation
- High precision
NOTE: Until 10th October 2017 it is still allowed to use mercury thermometers, if they are exclusively intended to be used for tests as per standards, which require the usage of mercury thermometers.
Certifications: ASTM
Specifications
- Beskrivelse:ASTM hydrometer
- Længde:330 mm
- Seq:001
- Område:19…31 °API