Bent forceps
Curved, pointed, 105 mm
Katalog # 232-0130
Gå til:
Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
FORCEPS 18/10 105MM POINTED STR STD 1 * 1 Items Katalog #232-2121
Product 232-2121 is similar to 232-0130 but manufacturer and specifications may vary.
Product Details & Documents
Stainless steel 18/8.
- Pointed, curved
- With serrated grip and tip
Specifikationer
- Model:Curved, pointed
- Længde:105 mm